Skip to Content
Merck
All Photos(1)

Key Documents

370444

Sigma-Aldrich

Dimethyl 2,3-indoledicarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO4
CAS Number:
Molecular Weight:
233.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

113-114 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1[nH]c2ccccc2c1C(=O)OC

InChI

1S/C12H11NO4/c1-16-11(14)9-7-5-3-4-6-8(7)13-10(9)12(15)17-2/h3-6,13H,1-2H3

InChI key

JNRXSMOJWKDVHC-UHFFFAOYSA-N

General description

Dimethyl 2,3-indoledicarboxylate is an heterocyclic building block.

Application

Reactant for preparation of:
  • Indole-indolone scaffolds
  • Chemiluminescent agents
  • Antihypertensive agents
  • Inhibitors of blood platelet aggregation and inotropic agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service