Skip to Content
Merck
All Photos(2)

Key Documents

195936

Sigma-Aldrich

3-(Perfluorobutyryl)-(+)-camphor

96%

Synonym(s):

(+)-3-(Heptafluorobutyryl)-(+)-camphor, 3-(Perfluorobutyryl)-D-camphor

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$193.00

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$193.00

About This Item

Empirical Formula (Hill Notation):
C14H15F7O2
CAS Number:
Molecular Weight:
348.26
Beilstein:
2306496
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

optical activity

[α]20/D +125°, c = 2.6 in chloroform

refractive index

n20/D 1.421 (lit.)

bp

60-70 °C/0.2 mmHg (lit.)

density

1.218 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC1(C)C2CC[C@@]1(C)C(=O)C2C(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C14H15F7O2/c1-10(2)6-4-5-11(10,3)8(22)7(6)9(23)12(15,16)13(17,18)14(19,20)21/h6-7H,4-5H2,1-3H3/t6-,7?,11+/m1/s1

InChI key

PEWOESYEGLBLNR-XGLFCGLISA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Perfluorobutyryl)-(+)-camphor can be used:
  • As a ligand in the synthesis of chiral europium complex for optoelectronic and photonic applications.[1]
  • As a ligand in the preparation of sodium tetrakis(3-heptafluorobutylryl-(+)-camphorato) Ln(III) complexes applicable as chemical sensors and NMR shift reagents.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Configurational Structures of Sodium Tetrakis (3-heptafluorobutylryl-(+)-camphorato) Ln (III) Complexes with a Trapped Na+ by Na+?FC Interactions in the Solid State and in Solution
Shirotani D, et al.
Inorganic Chemistry, 45(16), 6111-6113 (2006)
Highly circularly polarized electroluminescence from a chiral europium complex
Zinna F, et al.
Advanced Materials, 27(10), 1791-1795 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service