Skip to Content
Merck
All Photos(2)

Key Documents

189006

Sigma-Aldrich

(+)-3-(Trifluoroacetyl)camphor

98%

Synonym(s):

3-(Trifluoroacetyl)-D-camphor

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$102.00

$102.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$102.00

About This Item

Empirical Formula (Hill Notation):
C12H15F3O2
CAS Number:
Molecular Weight:
248.24
Beilstein:
6779716
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

$102.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

98%

form

liquid

optical activity

[α]19/D +148°, c = 2.3 in methylene chloride

refractive index

n20/D 1.451 (lit.)

bp

100-101 °C/16 mmHg (lit.)

density

1.172 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC1(C)C2CC[C@@]1(C)C(=O)C2C(=O)C(F)(F)F

InChI

1S/C12H15F3O2/c1-10(2)6-4-5-11(10,3)8(16)7(6)9(17)12(13,14)15/h6-7H,4-5H2,1-3H3/t6-,7?,11+/m1/s1

InChI key

ISLOIHOAZDSEAJ-XGLFCGLISA-N

Looking for similar products? Visit Product Comparison Guide

Application

(+)-3-(Trifluoroacetyl)camphor reacts with suitable chiral ketone precursors to generate chiral dioxiranes in situ, which can catalyse the direct asymmetric epoxidation of unfunctionalized olefins.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective epoxidation of unfunctionalized alkenes using dioxiranes generated in situ.
Curci R, et al.
Tetrahedron Letters, 36(32), 5831-5834 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service