Skip to Content
Merck
All Photos(1)

Key Documents

900264

Sigma-Aldrich

(−)-Sparteine

Synonym(s):

(−)-Lupinidine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$187.00

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$187.00

About This Item

Empirical Formula (Hill Notation):
C15H26N2
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.528 (lit.)

bp

137-138 °C/1 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@H]34)[C@@H]2C1

InChI

1S/C15H26N2/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17/h12-15H,1-11H2/t12-,13-,14-,15+/m0/s1

InChI key

SLRCCWJSBJZJBV-ZQDZILKHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service