Skip to Content
Merck
All Photos(1)

Key Documents

335037

Sigma-Aldrich

Dihydridotetrakis(triphenylphosphine)ruthenium(II)

Synonym(s):

Tetrakis(triphenylphosphine)ruthenium dihydride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$186.00

$186.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$186.00

About This Item

Linear Formula:
[(C6H5)3P]4RuH2
CAS Number:
Molecular Weight:
1156.26
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$186.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst

mp

181-183 °C (lit.)

SMILES string

[RuH2].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Ru.2H/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h4*1-15H;;;

InChI key

RXFATFKBDIQXLS-UHFFFAOYSA-N

Application

Catalyst for hydration of nitriles, isomerization of alkynes,Knoevenagel reactions and conjugate additions, and transferhydrogenation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naota, T.; Taki, H. et al.
Journal of the American Chemical Society, 111, 5954-5954 (1989)
Murahashi, S-I.; Sasao, S. et al.
The Journal of Organic Chemistry, 57, 2521-2521 (1992)
Ma, D.; Lu, X.
Tetrahedron Letters, 30, 843-843 (1989)

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service