Skip to Content
Merck
All Photos(1)

Key Documents

334480

Sigma-Aldrich

Iridium(IV) chloride hydrate

technical grade

Synonym(s):

Iridium tetrachloride hydrate, Tetrachloroiridium hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$210.00

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$210.00

About This Item

Linear Formula:
lrCl4 · xH2O
CAS Number:
Molecular Weight:
334.03 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
crystalline

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

form

crystalline

reaction suitability

reagent type: catalyst
core: iridium

concentration

48-55% Ir (after reduction with magnesium, gravimetric)

mp

>350 °C (lit.)

SMILES string

O.Cl[Ir](Cl)(Cl)Cl

InChI

1S/4ClH.Ir.H2O/h4*1H;;1H2/q;;;;+4;/p-4

InChI key

KVDBPOWBLLYZRG-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service