Skip to Content
Merck
All Photos(1)

Key Documents

203491

Sigma-Aldrich

Iridium(III) chloride hydrate

99.9% trace metals basis

Synonym(s):

Iridium trichloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$450.00

$450.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$450.00

About This Item

Linear Formula:
IrCl3 · xH2O
CAS Number:
Molecular Weight:
298.58 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

$450.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: iridium

impurities

≤1500.0 ppm Trace Metal Analysis

density

5.3 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].Cl[Ir](Cl)Cl

InChI

1S/3ClH.Ir.H2O/h3*1H;;1H2/q;;;+3;/p-3

InChI key

MJRFDVWKTFJAPF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iridium(III) chloride hydrate is a dark green to black hygroscopic solid that is widely used as a Ir source/precursor to synthesize Ir nanoparticle and battery materials.

Application

Iridium(III) chloride hydrate can be used:
  • As a precursor to synthesize IrO2 nanofibrous catalyst for oxygen evolution reaction.
  • As a starting material to prepare Ir complex that acts as an additive to hole transport layer in perovskite solar cells. This Ir complex helps to reduce recombination rate and enhances the stability of solar cells.
  • To fabricate thin and highly active IrO2 anodes for proton exchange membrane electrolyzer cells (PEMECs).
  • As a precursor to synthesize ionic Ir metal complexes for efficient flexible green light-emitting electrochemical cells. This leads to enhanced quantum efficiency and luminescence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaped Ir?Ni bimetallic nanoparticles for minimizing Ir utilization in oxygen evolution reaction.
Lim J,et al.
Chemical Communications (Cambridge, England), 52(32), 5641-5644 (2016)
Maximilian Moser et al.
Angewandte Chemie (International ed. in English), 53(33), 8628-8633 (2014-06-04)
Rutile TiO2 is a heavily investigated oxide with, to date, scarce applications in industrial catalysis. The inactivity of this material in oxidations has been related to its inability to dissociate molecular oxygen. Herein we show how rutile catalyzes the oxidation
Ann M Rajnicek et al.
Advanced healthcare materials, 7(17), e1800473-e1800473 (2018-07-06)
Innovative neurostimulation therapies require improved electrode materials, such as poly(3,4-ethylenedioxythiophene) (PEDOT) polymers or IrOx mixed ionic-electronic conductors and better understanding of how their electrochemistry influences nerve growth. Amphibian neurons growing on transparent films of electronic (metal) conductors and electronic-ionic conductors
Ultrabright oxygen optodes based on cyclometalated iridium (III) coumarin complexes.
Borisov SM and Klimant I
Analytical Chemistry, 79(19), 7501-7509 (2007)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service