Skip to Content
Merck
All Photos(1)

Key Documents

208973

Sigma-Aldrich

Hydrogen hexachloroiridate(IV) hydrate

Synonym(s):

Dihydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Hydrogen iridium hexachloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$289.00
5 G
$829.00

$289.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$289.00
5 G
$829.00

About This Item

Linear Formula:
H2Cl6Ir · xH2O
CAS Number:
Molecular Weight:
406.95 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
(Powder or Crystals or Granules or Beads)

$289.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

form

(Powder or Crystals or Granules or Beads)

Quality Level

concentration

36.0-44.0% Ir (gravimetric)

mp

65 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].[H]O[H].Cl[Ir](Cl)(Cl)Cl

InChI

1S/6ClH.Ir.H2O/h6*1H;;1H2/q;;;;;;+4;/p-4

InChI key

YDVQBPXDKJKDME-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrogen hexachloroiridate(IV) hydrate is black crystalline solid that is soluble in water. It is a coordination complex of iridium in the +4 oxidation state. It widely used as a precursor to prepare Ir based catalysts for various electrochemical applications.

Application

Hydrogen hexachloroiridate(IV) hydrate can be used:
  • To fabricate dimensionally stable electrodes for oxygen evolution reaction. It allows for the controlled deposition of iridium oxide, which is crucial for achieving stable and efficient electrodes for the OER.
  • As a precursor to prepare electroplating solution to modify ultra flexible neural electrodes. It helps to achieve high charge storage capacity and working stability.
  • As a Ir source to prepare Ir nanodendrite water splitting catalyst with large surface area and high electrochemical porosity.

Features and Benefits

Catalyzes the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes. Utilized in the formation of iridium-substituted Dawson- and Keggin-type polyoxometallates by refilling the vacant site of the lacunary precursors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic insights into the effect of promoters on Co/Al2O3 for Fischer-Tropsch synthesis
Xiaoli Yang, et al.
Chemical Engineering Journal, 445, 136655-136655 (2022)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service