Skip to Content
Merck
All Photos(2)

Key Documents

455962

Sigma-Aldrich

Hydrogen hexachloroiridate(IV) hydrate

99.9% trace metals basis

Synonym(s):

Dihydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Hydrogen iridium hexachloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$357.00

$357.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$357.00

About This Item

Linear Formula:
H2Cl6Ir · xH2O
CAS Number:
Molecular Weight:
406.95 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

$357.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

99.9% trace metals basis

form

solid

impurities

≤550.0 ppm Trace Metal Analysis

mp

65 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].[H]O[H].Cl[Ir](Cl)(Cl)Cl

InChI

1S/6ClH.Ir.H2O/h6*1H;;1H2/q;;;;;;+4;/p-4

InChI key

YDVQBPXDKJKDME-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Boron Nitride: A Support to Synthesize Stable Pt-Based Catalysts for High-Temperature Oxidative Reactions: This study discusses the use of hydrogen hexachloroiridate(IV) hydrate for stabilizing platinum-based catalysts on boron nitride supports, highlighting its importance in high-temperature catalytic applications (FBA Rahman, W Diao, JM Tengco, 2019).
  • Iridium–Palladium Binary Alloy as a Counter Electrode in Dye-Sensitized Solar Cells: This study investigates the effects of hydrogen hexachloroiridate(IV) hydrate on the properties of iridium-palladium binary alloys used as counter electrodes in solar cells (NAS Aziz, MYA Rahman, AA Umar, 2023).

Features and Benefits

Catalyzes the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes. Utilized in the formation of iridium-substituted Dawson- and Keggin-type polyoxometallates by refilling the vacant site of the lacunary precursors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service