Skip to Content
Merck
All Photos(1)

Key Documents

326208

Sigma-Aldrich

2-(Diphenylphosphino)benzaldehyde

97%

Synonym(s):

(2-Formylphenyl)diphenylphosphine, o-(Diphenylphosphino)benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00

$176.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$176.00

About This Item

Linear Formula:
(C6H5)2PC6H4CHO
CAS Number:
Molecular Weight:
290.30
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

$176.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

112-115 °C (lit.)

functional group

aldehyde
phosphine

SMILES string

O=Cc1ccccc1P(c2ccccc2)c3ccccc3

InChI

1S/C19H15OP/c20-15-16-9-7-8-14-19(16)21(17-10-3-1-4-11-17)18-12-5-2-6-13-18/h1-15H

InChI key

DRCPJRZHAJMWOU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Božidar Čobeljić et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 21(2), 145-162 (2015-11-28)
Square-planar azido Ni(II) complex with condensation product of 2-(diphenylphosphino)benzaldehyde and Girard's T reagent was synthesized and its crystal structure was determined. Cytotoxic activity of the azido complex and previously synthesized isothiocyanato, cyanato and chlorido Ni(II) complexes with this ligand was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service