Skip to Content
Merck
All Photos(1)

Key Documents

716626

Sigma-Aldrich

2-(Di-p-tolylphosphino)benzaldehyde

97%

Synonym(s):

2-[Bis(4-methylphenyl)phosphino]benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$154.00

$154.00


Estimated to ship onMarch 12, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$154.00

About This Item

Empirical Formula (Hill Notation):
C21H19OP
CAS Number:
Molecular Weight:
318.35
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

$154.00


Estimated to ship onMarch 12, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

105-111 °C

functional group

aldehyde
phosphine

SMILES string

Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccccc3C=O

InChI

1S/C21H19OP/c1-16-7-11-19(12-8-16)23(20-13-9-17(2)10-14-20)21-6-4-3-5-18(21)15-22/h3-15H,1-2H3

InChI key

IDDRQJVTYBKJLP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service