Skip to Content
Merck
All Photos(2)

Documents

A65204

Sigma-Aldrich

2-Picolylamine

99%

Synonym(s):

2-(Aminomethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
108054
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

82-85 °C/12 mmHg (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

NCc1ccccn1

InChI

1S/C6H8N2/c7-5-6-3-1-2-4-8-6/h1-4H,5,7H2

InChI key

WOXFMYVTSLAQMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Picolylamine is a bihaptic amine that can be used:
  • As a key precursor to synthesize various ionic liquids through the formation of β-amino alcohols as intermediates.
  • As a chelating ligand for the synthesis of complexes such as zinc picolylamine complex and Cu(II) picolylamine complex.
  • To functionalize poly(styrene-co-maleic anhydride) (PSMA) resin to facilitate the adsorption of uranium from aqueous solution.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Structure of tris(2-picolylamine) zinc (II) dichloride-ethanol,[Zn (α-pic)3] Cl2.EtOH; a complex related to an FeII spin crossover complex.
Mikami-Kido M and Saito Y
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 38(2), 452-455 (1982)
Recovery of uranium (VI) from aqueous solution by 2-picolylamine functionalized poly (styrene-co-maleic anhydride) resin.
Liu S, et al.
Journal of Colloid and Interface Science, 497(16), 385-392 (2017)
Glycosylated metal chelators as anti-parasitic agents with tunable selectivity.
Reddy A, et al.
Dalton Transactions, 46(16), 5297-5307 (2017)
Cengiz Yenikaya et al.
Bioorganic & medicinal chemistry, 18(2), 930-938 (2009-12-17)
A novel proton transfer compound, pyridin-2-ylmethanaminium 2,4-dichloro-5-sulfamoylbenzoate (1), and a mixed-ligand Zn(II) complex, bis(2,4-dichloro-5-sulfamoylbenzoate)(2-aminomethylpyridine)aquazinc(II) monohydrate (2), have been synthesized from the same free ligands, which are 2,4-dichloro-5-sulfamoylbenzoic acid (Hsba) and 2-aminomethylpyridine (amp). They have been characterized by elemental, spectral ((1)H
Solvent-free Al(OTf)3-catalyzed aminolysis of 1, 2-epoxides by 2-picolylamine: a key step in the synthesis of ionic liquids.
Fringuelli F, et al.
The Journal of Organic Chemistry, 69(22), 7745-7747 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service