Skip to Content
Merck
All Photos(1)

Key Documents

43163

Sigma-Aldrich

3-(Diphenylphosphino)-1-propylamine

technical, ≥90% (GC)

Synonym(s):

(3-Aminopropyl)diphenylphosphine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$322.00

$322.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$322.00

About This Item

Empirical Formula (Hill Notation):
C15H18NP
CAS Number:
Molecular Weight:
243.28
Beilstein:
2730850
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$322.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

grade

technical

Assay

≥90% (GC)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

functional group

phosphine

SMILES string

NCCCP(c1ccccc1)c2ccccc2

InChI

1S/C15H18NP/c16-12-7-13-17(14-8-3-1-4-9-14)15-10-5-2-6-11-15/h1-6,8-11H,7,12-13,16H2

InChI key

DQZWMOWSTWWMPP-UHFFFAOYSA-N

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Dellis et al.
BMC pharmacology, 11, 1-1 (2011-01-27)
Store-Operated Calcium Entry (SOCE) is the major Ca2+ ion entry pathway in lymphocytes and is responsible of a severe combined immunodeficiency (SCID) when deficient. It has recently been observed or highlighted in other cell types such as myoblasts and neurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service