Skip to Content
Merck
All Photos(1)

Key Documents

298336

Sigma-Aldrich

3-(Perfluorobutyryl)-(−)-camphor

98%

Synonym(s):

3-(Heptafluorobutyryl)-L-camphor, 3-Heptafluorobutyryl-(−)-camphor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15F7O2
CAS Number:
Molecular Weight:
348.26
Beilstein:
2306496
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]19/D −123°, c = 2.6 in chloroform

refractive index

n20/D 1.422 (lit.)

density

1.218 g/mL at 25 °C (lit.)

SMILES string

CC1(C)C2CC[C@]1(C)C(=O)C2C(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C14H15F7O2/c1-10(2)6-4-5-11(10,3)8(22)7(6)9(23)12(15,16)13(17,18)14(19,20)21/h6-7H,4-5H2,1-3H3/t6-,7?,11+/m0/s1

InChI key

PEWOESYEGLBLNR-NSMOOJLNSA-N

Application

3-(Perfluorobutyryl)-(−)-camphor can be used:
  • To prepare mesoporous metal-organic hybrid materials, which are used as catalysts in the asymmetric hetero-Diels-Alder cyclization of Danishefsky′s diene with benzaldehydes.
  • As a precursor in the synthesis of chiral tetrakis(β-diketonate) Ln(III) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening of rare earth metal grafted MCM-41 silica for asymmetric catalysis.
Gerstberger G and Anwander R.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 44, 303-310 (2001)
Yiji Lin et al.
Dalton transactions (Cambridge, England : 2003), 41(22), 6696-6706 (2012-04-28)
Chiral tetrakis(β-diketonate) Ln(III) complexes Δ-[NaLa(d-hfc)(4)(CH(3)CN)] (1) and Λ-[NaLa(l-hfc)(4) (CH(3)CN)] (2) (d/l-hfc(-) = 3-heptafluo-robutylryl-(+)/(-)-camphorate) are a pair of enantiomers and crystallize in the same Sohncke space group (P2(1)2(1)2(1)) with dodecahedral (DD) geometry. Typically positive and negative exciton splitting patterns around 320

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service