Skip to Content
Merck
All Photos(2)

Key Documents

176494

Sigma-Aldrich

Europium tris[3-(trifluoromethylhydroxymethylene)-(+)-camphorate]

Synonym(s):

Eu(facam)3, Eu(tfc)3, Europium(III) tris[3-(trifluoromethylhydroxymethylene)-d-camphorate], Tris(3-trifluoroacetyl-d-camphorato)europium(III), Tris[3-(trifluoromethylhydroxymethylene)-d-camphorato]europium(III)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$51.10
1 G
$172.00

$51.10


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$51.10
1 G
$172.00

About This Item

Empirical Formula (Hill Notation):
C36H42EuF9O6
CAS Number:
Molecular Weight:
893.66
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

$51.10


Estimated to ship onMay 28, 2025


Request a Bulk Order

optical activity

[α]20/D +165°, c = 1% in chloroform (solvent is ethanol free)

technique(s)

NMR: suitable

functional group

fluoro
ketone

SMILES string

CC1(C)C2CCC1(C)C(=O)\C2=C(/O[Eu](O\C(=C3\C4CCC(C)(C3=O)C4(C)C)C(F)(F)F)O\C(=C5\C6CCC(C)(C5=O)C6(C)C)C(F)(F)F)C(F)(F)F

InChI

1S/3C12H15F3O2.Eu/c3*1-10(2)6-4-5-11(10,3)8(16)7(6)9(17)12(13,14)15;/h3*6,17H,4-5H2,1-3H3;/q;;;+3/p-3/b3*9-7-;

InChI key

BWIHMSGCCRUUCZ-NCJHBDPTSA-K

Application

Optically active NMR shift reagent. Used with Ag(fod) for the enantiomeric resolution of sec-butylisothiouronium chloride.[1]
Reactant involved in:
  • The synthesis of europium acetylcamphorate phosphine oxide complexes
  • Reactions with 1,10-N,N′-phenanthroline dioxide derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Chemistry, 59, 562-562 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service