Skip to Content
Merck
All Photos(2)

Key Documents

296171

Sigma-Aldrich

Methyl 4-acetyl-5-oxohexanoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2CHCH2CH2CO2CH3
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.459 (lit.)

bp

182 °C/48 mmHg (lit.)

density

1.066 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

COC(=O)CCC(C(C)=O)C(C)=O

InChI

1S/C9H14O4/c1-6(10)8(7(2)11)4-5-9(12)13-3/h8H,4-5H2,1-3H3

InChI key

XFUDNZLAPUHONL-UHFFFAOYSA-N

Application

Methyl 4-acetyl-5-oxohexanoate is a β-ketoester, has been used:
  • in modified Knorr condensation for the synthesis of pyrrole[1]
  • to identify the Michael addition product by using GC and GC-MS[2]
  • in preparation of dibenzyl-3,6-dimethylpyrazine-2,5-dicarboxylate[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of a modified Knorr pyrrole condensation.
Rapoport H and Harbuct JW.
The Journal of Organic Chemistry, 36(6), 853-855 (1971)
Suppressed native hydrolytic activity of a lipase to reveal promiscuous Michael addition activity in water.
Svedendahl M, et al.
ChemCatChem, 1(2), 252-258 (2009)
C-H??? p and C= O??? p Intermolecular Interactions in Dibenzyl-3, 6-dimethylpyrazine-2, 5-dicarboxylate.
Silva JA, et al.
Journal of Chemical Crystallography, 38(4), 301-303 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service