Skip to Content
Merck
All Photos(1)

Key Documents

245054

Sigma-Aldrich

Methyltriphenylphosphonium iodide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$112.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$112.00

About This Item

Linear Formula:
CH3P(C6H5)3I
CAS Number:
Molecular Weight:
404.22
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

183-185 °C (lit.)

functional group

phosphine

SMILES string

[I-].C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1

InChI key

JNMIXMFEVJHFNY-UHFFFAOYSA-M

Application

Reactant for synthesis of:
  • Triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells
  • Diastereoisomeric carba analogs of glycal-derived vinyl epoxides for the production of carbasugars
  • Alkaloid natural products via organocascade catalysis
  • Triplet carbenes via oxidation of allene compounds
  • Stilbenes via dehydrohalogenation-Heck olefination and Wittig-Heck reactions

Legal Information

Product of Fluorous Technologies, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Mahdi Khataei et al.
Talanta, 178, 473-480 (2017-11-16)
In this study, a novel generation of deep eutectic solvents (DESs) was used as an acceptor phase in three-phase hollow fiber liquid phase microextraction (HF-LPME) based on two immiscible organic phases. It was compared with other common DESs for extraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service