Skip to Content
Merck
All Photos(1)

Key Documents

SML3598

Sigma-Aldrich

1PBC

≥98% (HPLC)

Synonym(s):

1-Hydroxy-3-(trifluoromethyl)pyrido[1,2-a]benzimidazole-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6F3N3O
CAS Number:
Molecular Weight:
277.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

OC1=CC(C(F)(F)F)=C(C#N)C2=NC3=CC=CC=C3N21

Biochem/physiol Actions

1PBC is a potent and selective blocker of TMEM16A calcium-activated chloride channel that interacts with the anion pore. Also, 1PBC is a potent inhibitor of TMEM16F channel.
Potent and selective blocker of TMEM16A calcium-activated chloride channel; potent inhibitor of TMEM16F channel

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a conserved drug binding pocket in TMEM16 proteins
Research square (2022)
Inhibition mechanism of the chloride channel TMEM16A by the pore blocker 1PBC
Lam AKM, Rutz S, Dutzler R
Nature Communications, 13, 2798-2798 (2022)
Christian J Peters et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(11), 3547-3552 (2015-03-04)
TMEM16A (transmembrane protein 16) (Anoctamin-1) forms a calcium-activated chloride channel (CaCC) that regulates a broad array of physiological properties in response to changes in intracellular calcium concentration. Although known to conduct anions according to the Eisenman type I selectivity sequence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service