Skip to Content
Merck
All Photos(1)

Key Documents

291366

Sigma-Aldrich

N-(2-Chloroethyl)dibenzylamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2NCH2CH2Cl · HCl
CAS Number:
Molecular Weight:
296.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

190-193 °C (lit.)

SMILES string

Cl.ClCCN(Cc1ccccc1)Cc2ccccc2

InChI

1S/C16H18ClN.ClH/c17-11-12-18(13-15-7-3-1-4-8-15)14-16-9-5-2-6-10-16;/h1-10H,11-14H2;1H

InChI key

LZXCEBPGNFLHEQ-UHFFFAOYSA-N

General description

N-(2-Chloroethyl)dibenzylamine hydrochloride is also referred as dibenamine. Mechanism of protection against carbon tetrachloride-induced hepatotoxicity produced by pretreatment with [N-(2-chloroethyl) dibenzylamine] hydrochloride has been investigated. It is a powerful adrenergic blocking agent, is known to modify the pharmacological effects of epinephrine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of dibenamine (N-(2-chloroethyl) dibenzylamine) on the metabolism of radioactive epinephrine.
R W SCHAYER et al.
The Journal of biological chemistry, 202(1), 39-43 (1953-05-01)
Nature of the protection against carbon tetrachloride-induced hepatotoxicity produced by pretreatment with dibenamine (N-(2-chloroethyl)dibenzylamine).
H M Maling et al.
Biochemical pharmacology, 23(10), 1479-1491 (1974-05-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service