Skip to Content
Merck
All Photos(1)

Key Documents

P32406

Sigma-Aldrich

3-Phenyl-1-propylamine

98%

Synonym(s):

3-Phenylpropylamine, γ-Aminopropylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$82.10
25 G
$121.00

$82.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$82.10
25 G
$121.00

About This Item

Linear Formula:
C6H5(CH2)3NH2
CAS Number:
Molecular Weight:
135.21
Beilstein:
2205526
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$82.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.524 (lit.)

bp

221 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

NCCCc1ccccc1

InChI

1S/C9H13N/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8,10H2

InChI key

LYUQWQRTDLVQGA-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Barros Almeida et al.
Cytokine, 131, 155100-155100 (2020-04-22)
There is growing evidence that there is a relationship between major depressive disorder (MDD), also simply known as "depression", and inflammatory processes. Selective serotonin inhibitors, such as fluoxetine, are used as a first-line treatment for depression, and it is hypothesized
Mohammad Darvishnejad et al.
Talanta, 199, 547-555 (2019-04-07)
In the present study, a low-cost, convenient and reliable thin film coating was developed as the extraction phase. The new sorbent was obtained by chemical bond formation between the hybrid sol-gel precursors and aluminum strip substrate in the first step
R Herráez-Hernández et al.
Journal of chromatography. B, Biomedical applications, 679(1-2), 69-78 (1996-04-26)
A method for the determination of amphetamine and related compounds in urine based on on-line derivatization with 9-fluorenylmethyl chloroformate (FMOC) and high-performance liquid chromatography is described. Derivatization is performed in a 20 x 2.1 mm I.D. column packed with a
Cyntia M Palacio et al.
The FEBS journal, 286(20), 4086-4102 (2019-06-05)
The biodegradation of the nylon-6 precursor caprolactam by a strain of Pseudomonas jessenii proceeds via ATP-dependent hydrolytic ring opening to 6-aminohexanoate. This non-natural ω-amino acid is converted to 6-oxohexanoic acid by an aminotransferase (PjAT) belonging to the fold type I pyridoxal
P F Fitzpatrick et al.
Archives of biochemistry and biophysics, 249(1), 70-75 (1986-08-15)
In order to determine the order of substrate binding to dopamine beta-hydroxylase during catalysis, the effect of alternate substrates upon kinetic parameters was examined. The V/K value for ascorbate was unchanged when tyramine, phenylpropylamine, p-Cl-phenethylamine, p-CH3O-phenethylamine, or phenethylamine was the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service