Skip to Content
Merck
All Photos(1)

Documents

H50004

Sigma-Aldrich

4-Hydroxyphenylacetic acid

98%

Synonym(s):

(4-Hydroxyphenyl)acetic acid, (p-Hydroxyphenyl)acetic acid, 2-(4′-Hydroxyphenyl)acetic acid, 2-[4-(Hydroxy)phenyl]acetic acid, 4-(Carboxymethyl)phenol, 4-Hydroxybenzeneacetic acid, 4′-Hydroxyphenylacetic acid, p-Hydroxybenzeneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
1448766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

148-151 °C (lit.)

SMILES string

OC(=O)Cc1ccc(O)cc1

InChI

1S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

XQXPVVBIMDBYFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Hydroxyphenylacetic acid is a phenolic acid that is esterified with various alcohols to synthesize esters.

Application

Reagent used in the acylation of phenols and amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of negative tone photoresist based on acid catalyzed dehydration crosslinking of novolac resins having pendant carboxyl groups
Liu HH, et al.
Journal of Polymer Research, 9, 251-256 (2002)
Journal of the Chemical Society. Perkin Transactions 1, 3005-3005 (1991)
Kenneth Stensrud et al.
The Journal of organic chemistry, 74(15), 5219-5227 (2009-07-04)
Three new trifluoromethylated p-hydroxyphenacyl (pHP)-caged gamma-aminobutyric acid (GABA) and glutamate (Glu) derivatives have been examined for their efficacy as photoremovable protecting groups in aqueous solution. Through the replacement of hydrogen with fluorine, e.g., a m-trifluoromethyl or a m-trifluoromethoxy versus m-methoxy
Korean J. Med. Chem., 1, 2-2 (1991)
Matthew Snelson et al.
Science advances, 7(14) (2021-04-02)
Intake of processed foods has increased markedly over the past decades, coinciding with increased microvascular diseases such as chronic kidney disease (CKD) and diabetes. Here, we show in rodent models that long-term consumption of a processed diet drives intestinal barrier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service