Skip to Content
Merck
All Photos(1)

Key Documents

C42602

Sigma-Aldrich

1-(2-Chloroethyl)piperidine hydrochloride

98%

Synonym(s):

2-Piperidinoethyl chloride hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$89.20

$89.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$89.20

About This Item

Empirical Formula (Hill Notation):
C7H14ClN · HCl
CAS Number:
Molecular Weight:
184.11
Beilstein:
3910268
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$89.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

mp

230-232 °C (lit.)

SMILES string

Cl[H].ClCCN1CCCCC1

InChI

1S/C7H14ClN.ClH/c8-4-7-9-5-2-1-3-6-9;/h1-7H2;1H

InChI key

VFLQQZCRHPIGJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Li-Na Zhu et al.
PloS one, 7(5), e35586-e35586 (2012-05-26)
The discovery of uncommon DNA structures and speculation about their potential functions in genes has brought attention to specific DNA structure recognition. G-quadruplexes are four-stranded nucleic acid structures formed by G-rich DNA (or RNA) sequences. G-rich sequences with a high
Eda Yuhana Ariffin et al.
Sensors (Basel, Switzerland), 18(4) (2018-04-13)
A sensitive and selective optical DNA biosensor was developed for dengue virus detection based on novel square-planar piperidine side chain-functionalized N,N'-bis-4-(hydroxysalicylidene)-phenylenediamine-nickel(II), which was able to intercalate via nucleobase stacking within DNA and be functionalized as an optical DNA hybridization marker.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service