Skip to Content
Merck
All Photos(1)

Key Documents

270776

Sigma-Aldrich

tert-Butyl formate

99%

Synonym(s):

Formic acid tert-butylester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$160.00
25 G
$461.00

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$160.00
25 G
$461.00

About This Item

Linear Formula:
HCOOC(CH3)3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1738871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.379 (lit.)

bp

82-83 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC=O

InChI

1S/C5H10O2/c1-5(2,3)7-4-6/h4H,1-3H3

InChI key

RUPAXCPQAAOIPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl formate is an organic chemical compound obtained during the decomposition of methyl tert-butyl ether (MTBE).

Application

tert-Butyl formate can be used as a reagent in the direct carbonylation of benzyl halides in the presence of a rhodium catalyst.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K F Chen et al.
Water environment research : a research publication of the Water Environment Federation, 81(7), 687-694 (2009-08-21)
The objective of this study was to evaluate the feasibility of using ferrous ion-activated persulfate oxidation to remediate groundwater contaminated with methyl tert-butyl ether (MTBE). In this study, batch experiments were conducted to evaluate the effects of various factors on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service