Skip to Content
Merck
All Photos(3)

Key Documents

689890

Sigma-Aldrich

(R)-3,3′-Bis(2,4,6-triisopropylphenyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate

greener alternative

≥97.0% (qNMR)

Synonym(s):

(11bR)-4-Hydroxy-2,6-bis[2,4,6-tris(1-methylethyl)phenyl]dinaphtho[2,1-d:1′,2′-f]-1,3,2-dioxaphosphepin 4-oxide, (R)-3,3′-Bis(2,4,6-triisopropylphenyl)-1,1′-bi-2-naphthol cyclic monophosphate, (R)-TRIP

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$391.00
500 MG
$1,840.00

$391.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$391.00
500 MG
$1,840.00

About This Item

Empirical Formula (Hill Notation):
C50H57O4P
CAS Number:
Molecular Weight:
752.96
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$391.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (qNMR)

form

lumps

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

functional group

phosphate

greener alternative category

SMILES string

CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c2cc3ccccc3c-4c2OP(O)(=O)Oc5c(cc6ccccc6c-45)-c7c(cc(cc7C(C)C)C(C)C)C(C)C

InChI

1S/C50H57O4P/c1-27(2)35-23-39(29(5)6)45(40(24-35)30(7)8)43-21-33-17-13-15-19-37(33)47-48-38-20-16-14-18-34(38)22-44(50(48)54-55(51,52)53-49(43)47)46-41(31(9)10)25-36(28(3)4)26-42(46)32(11)12/h13-32H,1-12H3,(H,51,52)

InChI key

AGQAQYPGJDBIQR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Versatile organocatalyst. Uses include asymmetric reductive amination, enantioselective Diels-Alder reactions and asymmetric counteranion-directed catalysis (ACDC). Can be used in cooperation with chiral iridium complexes to catalyze greener asymmetric reductive amination using hydrogen gas.

Metal-Brønsted Acid Cooperative Catalysis for Asymmetric Reductive Amination

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Related Content

The List group focuses on the development of new concepts in catalysis. Since 1999, this research group has pioneered the development of organocatalysis as the third pillar of stereoselective catalysis, along side biocatalysis and transition metal catalysis.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service