Skip to Content
Merck
All Photos(2)

Key Documents

261521

Sigma-Aldrich

Butyl formate

97%

Synonym(s):

Formic acid butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$88.80

$88.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$88.80

About This Item

Linear Formula:
HCOO(CH2)3CH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1742108
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.389 (lit.)

bp

106-107 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CCCCOC=O

InChI

1S/C5H10O2/c1-2-3-4-7-5-6/h5H,2-4H2,1H3

InChI key

NMJJFJNHVMGPGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Absolute rate coefficient for the reaction of Cl atom with butyl formate has been measured by laser photolysis-resonance fluorescence technique[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Absolute rate constants for the reactions of Cl atoms with a series of esters.
Notario A, et al.
The Journal of Physical Chemistry A, 102(18), 3112-3117 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service