Skip to Content
Merck
All Photos(1)

Key Documents

674605

Sigma-Aldrich

(R)-3,3′-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate

95%

Synonym(s):

(11bR)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-hydroxydinaphtho[2,1-d:1′,2′-f]-1,3,2-dioxaphosphepin 4-oxide, (R)-3,3′-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1′-bi-2-naphthol cyclic monophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$246.00

$246.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$246.00

About This Item

Empirical Formula (Hill Notation):
C36H17F12O4P
CAS Number:
Molecular Weight:
772.47
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$246.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

optical activity

[α]20/D -220°, c = 1 in chloroform (typical)

mp

162-175 °C

SMILES string

OP1(=O)Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)-c6cc(cc(c6)C(F)(F)F)C(F)(F)F)-c7cc(cc(c7)C(F)(F)F)C(F)(F)F

InChI

1S/C36H17F12O4P/c37-33(38,39)21-9-19(10-22(15-21)34(40,41)42)27-13-17-5-1-3-7-25(17)29-30-26-8-4-2-6-18(26)14-28(32(30)52-53(49,50)51-31(27)29)20-11-23(35(43,44)45)16-24(12-20)36(46,47)48/h1-16H,(H,49,50)

InChI key

DQORDVSQWPKAQJ-UHFFFAOYSA-N

General description

(R)-3,3′-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate is a BINOL-based chiral phosphoric acid, which can efficiently catalyze the nucleophilic addition to imine substrates. [BINOL=1,1′-bi-2-naphthol]

Application

As a chiral Bronsted acid, BINOL- derived cyclic phosphoric acid (such as Akiyama chiral Bronsted acid) was also shown to be an efficient catalyst for the hydrophosphonylation of aldimines at room temperature.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiko Akiyama et al.
Organic letters, 7(13), 2583-2585 (2005-06-17)
[reaction: see text] A cyclic phosphoric acid derivative, derived from (R)-BINOL, was used as a chiral Brønsted acid (10 mol %) in hydrophosphonylation of aldimines with diisopropyl phosphite at room temperature. Alpha-amino phosphonates were obtained with good to high enantioselectivities.

Articles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service