Skip to Content
Merck
All Photos(1)

Key Documents

269948

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine nickel(II)

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$212.00

$212.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$212.00

About This Item

Empirical Formula (Hill Notation):
C36H44N4Ni
CAS Number:
Molecular Weight:
591.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$212.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst
core: nickel

λmax

392 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Ni]n34)c(CC)c5CC

InChI

1S/C36H44N4.Ni/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;/h17-20H,9-16H2,1-8H3;/q-2;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;

InChI key

DIGQQRGHPATISA-XTPDIVBZSA-N

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine nickel can be formed by metalating 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine with nickel (II).1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service