Skip to Content
Merck
All Photos(1)

Key Documents

392448

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine ruthenium(II) carbonyl

Dye content ~80 %

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$298.00

$298.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$298.00

About This Item

Empirical Formula (Hill Notation):
C45H28N4ORu
CAS Number:
Molecular Weight:
741.80
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$298.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

composition

Dye content, ~80%

reaction suitability

reagent type: catalyst
core: ruthenium

λmax

410 nm

SMILES string

[C-]#[O+].[Ru]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.CO.Ru/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;1-2;/h1-28H;;/q-2;;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;

InChI key

AALUWVZJQWOFBD-YKKPBKTHSA-N

Other Notes

may contain solvent of crystallization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service