Skip to Content
Merck
All Photos(1)

Key Documents

257532

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine iron(III) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$191.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C36H44ClFeN4
CAS Number:
Molecular Weight:
624.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

λmax

382 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Fe](Cl)n34)c(CC)c5CC

InChI

1S/C36H44N4.ClH.Fe/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;;/h17-20H,9-16H2,1-8H3;1H;/q-2;;+3/p-1/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;;

InChI key

NWWCYXFDLUMYRQ-YAJYDHHFSA-M

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine metalates with iron chloride to form 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine iron chloride (FeOEP). It may be used to form Langmuir Blodgett (LB) thin films. Iron–porphyrin could be present naturally in biofilms.[1]

Application

An iron–porphyrin adlayer adsorbed on the Au(111) substrate was used to study the electrocatalytic reduction of O2 in HClO4.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Situ Scanning Tunneling Microscopy Study of the Effect of Iron Octaethylporphyrin Adlayer on the Electrocatalytic Reduction of O2 on Au (111)
Yoshimoto S, et al.
The Journal of Physical Chemistry B, 108(17), 5171-5174 (2044)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service