Skip to Content
Merck
All Photos(3)

Key Documents

78239

Sigma-Aldrich

(R)-3-Phenylbutyric acid

≥98.5% (sum of enantiomers, GC)

Synonym(s):

(R)-β-Methylhydrocinnamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$186.00

$186.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$186.00

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein:
2501477
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$186.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Assay

≥98.5% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −57±2°, c = 1% in benzene

refractive index

n20/D 1.517

bp

94-95 °C/0.3 mmHg (lit.)

density

1.069 g/mL at 20 °C (lit.)

functional group

carboxylic acid
phenyl

SMILES string

C[C@H](CC(O)=O)c1ccccc1

InChI

1S/C10H12O2/c1-8(7-10(11)12)9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,11,12)/t8-/m1/s1

InChI key

ZZEWMYILWXCRHZ-MRVPVSSYSA-N

Other Notes

Chiral standard. Chiral building block for the synthesis of the corresponding alcohol, amide and olefin[1][2][3]; Stereoselective α-amination via Evans′ imide enolate[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Dharanipragada et al.
Tetrahedron Letters, 30, 6841-6841 (1989)
T. Kogure et al.
The Journal of Organic Chemistry, 49, 576-576 (1984)
D.J. Cram
Journal of the American Chemical Society, 74, 2137-2137 (1952)
R. Bussas et al.
The Journal of Organic Chemistry, 48, 2828-2828 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service