Skip to Content
Merck
All Photos(4)

Key Documents

232785

Sigma-Aldrich

Tungsten(VI) oxide

powder, ≤25 μm, ≥99% trace metals basis

Synonym(s):

Tungstic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$256.00
500 G
$868.00
2 KG
$2,280.00

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$256.00
500 G
$868.00
2 KG
$2,280.00

About This Item

Linear Formula:
WO3
CAS Number:
Molecular Weight:
231.84
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99% trace metals basis

form

powder

particle size

≤25 μm

density

7.16 g/mL at 25 °C (lit.)

SMILES string

O=[W](=O)=O

InChI

1S/3O.W

InChI key

ZNOKGRXACCSDPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tungsten(VI) oxide, also known as tungsten trioxide, is a chemical compound with the formula WO3. Our material is a yellow crystalline solid. It is a versatile material with applications in electrochromic devices, sensors, photocatalysis, energy storage, and various other fields in material science. WO3 is used in gas and humidity sensors due to its sensitivity to various gases. It is also used as a starting material for the synthesis of tungstates, which have applications in x-ray screen phosphors and mineral separation.

Application


  • Tuning the Photo-Luminescence Properties of WO(3) Layers by the Adjustment of Layer Formation Conditions.: This research explores the photo-luminescence properties of tungsten(VI) oxide (WO3) layers and how they can be adjusted by varying the layer formation conditions. Such tuning of properties can have significant applications in energy storage and battery materials due to the enhanced electrochemical properties that can be achieved (Petruleviciene et al., 2020).

  • Decomposition of Formic Acid Using Tungsten(VI) Oxide Supported AgPd Nanoparticles.: This study investigates the catalytic properties of tungsten(VI) oxide (WO3) supported silver-palladium (AgPd) nanoparticles in the decomposition of formic acid. The findings highlight the potential industrial applications of WO3 in chemical reactions and energy storage systems (Akbayrak, 2019).


Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Questions

  1. Hallo, färbt sich das Wolframtrioxid blau wenn man es im Wasser löst und mit glühender kohle bearbeitet ?

    1 answer
    1. Tungsten(VI) oxide (WO3) does not turn blue when treated with glowing charcoal. WO3 is generally water-insoluble. While WO3 itself is generally yellow, the reduction of WO3 by carbon may lead to the formation of different tungsten oxides (like WO3), which can exhibit various colors. However, the direct process described does not typically produce a blue solution. The resulting color changes depend on the specific reduction products and their oxidation states.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service