Skip to Content
Merck
All Photos(1)

Key Documents

400505

Sigma-Aldrich

Tungsten(IV) oxide

−100 mesh, 99.99% trace metals basis

Synonym(s):

Tungsten dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$154.00

About This Item

Linear Formula:
WO2
CAS Number:
Molecular Weight:
215.84
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

particle size

−100 mesh

SMILES string

O=[W]=O

InChI

1S/2O.W

InChI key

DZKDPOPGYFUOGI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gil Sharon et al.
Cell, 177(6), 1600-1618 (2019-06-01)
Autism spectrum disorder (ASD) manifests as alterations in complex human behaviors including social communication and stereotypies. In addition to genetic risks, the gut microbiome differs between typically developing (TD) and ASD individuals, though it remains unclear whether the microbiome contributes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service