Skip to Content
Merck
All Photos(1)

Key Documents

196061

Sigma-Aldrich

4-Methoxy-3-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$69.20

$69.20


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$69.20

About This Item

Linear Formula:
CH3OC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
197.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.20


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

98%

mp

192-194 °C (lit.)

SMILES string

COc1ccc(cc1[N+]([O-])=O)C(O)=O

InChI

1S/C8H7NO5/c1-14-7-3-2-5(8(10)11)4-6(7)9(12)13/h2-4H,1H3,(H,10,11)

InChI key

ANXBDAFDZSXOPQ-UHFFFAOYSA-N

Application

4-Methoxy-3-nitrobenzoic acid was used in the synthesis of:
  • BIPHEP-1-OMe [BIHEP= (2,2′-bis(diphenylphosphino)biphenyl)], precatalyst for reductive coupling of butadiene to aldehydes[1]
  • aniline mustard analogues, having potential anti-tumor activity[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason R Zbieg et al.
Advanced synthesis & catalysis, 352(14-15), 2416-2420 (2010-12-18)
Exposure of alcohols 1a-1i to butadiene in the presence of a cyclometallated iridium catalyzed derived from allyl acetate, 4-methoxy-3-nitrobenzoic acid and BIPHEP (2,2'-bis(diphenylphosphino)biphenyl) results in hydrogen transfer to generate aldehyde-allyliridium pairs, which engage in C-C coupling to form products of
B G Choi et al.
Archives of pharmacal research, 21(2), 157-163 (1999-01-06)
Several aniline mustard analogues were obtained by introducing N,N-bis(2-chloroethyl)amino moiety to phenyl ring of A10 analogues in order to increase reactivity of A10 analogs and selectivity into DNA. The in vitro antitumor activity of synthesized compounds was evaluated using five

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service