Skip to Content
Merck
All Photos(3)

Key Documents

204781

Sigma-Aldrich

Tungsten(VI) oxide

powder, 99.995% trace metals basis

Synonym(s):

Tungstic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$205.00
50 G
$634.00

$205.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$205.00
50 G
$634.00

About This Item

Linear Formula:
WO3
CAS Number:
Molecular Weight:
231.84
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38180904
PubChem Substance ID:
NACRES:
NA.23

$205.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.995% trace metals basis

form

powder

density

7.16 g/mL at 25 °C (lit.)

SMILES string

O=[W](=O)=O

InChI

1S/3O.W

InChI key

ZNOKGRXACCSDPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tungsten(VI) oxide is an n-type oxygen deficient semiconducting metal oxide widely used to fabricate gas sensing devices.

It can also be used to prepare double perovskite materials for variousapplications.

Application

  • Coordination complexes of the tungsten (VI) oxide fluorides WOF4 and WO2F2 with neutral oxygen-and nitrogen-donor ligands: This study explores the coordination chemistry of tungsten(VI) oxide fluorides, which could be significant for researchers interested in the reactivity and applications of tungsten complexes (Levason et al., 2016).
  • Aerosol-assisted chemical vapor deposition of tungsten oxide films and nanorods from oxo tungsten (VI) fluoroalkoxide precursors: The study explores the deposition processes for creating tungsten oxide films and nanorods, relevant for applications in electronics and nanotechnology (Kim et al., 2015).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franco Dinelli et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-17)
In this paper, we present a study of tungsten disulfide (WS2) two-dimensional (2D) crystals, grown on epitaxial Graphene. In particular, we have employed scanning electron microscopy (SEM) and µRaman spectroscopy combined with multifunctional scanning probe microscopy (SPM), operating in peak

Articles

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Questions

  1. Guten Tag, ich möchte gerne wissen, wie groß die Pulver sind?

    1 answer
    1. The particle size of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service