Skip to Content
Merck
All Photos(3)

Key Documents

515973

Sigma-Aldrich

Potassium hexafluorophosphate

99.5% trace metals basis

Synonym(s):

Potassium fluophosphate, Potassium hexafluorophosphate, Potassium hexafluorophosphate (KPF)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$61.90
25 G
$220.00

$61.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$61.90
25 G
$220.00

About This Item

Linear Formula:
KPF6
CAS Number:
Molecular Weight:
184.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

$61.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99.5% trace metals basis

form

solid

impurities

≤5500.0 ppm Trace Metal Analysis

mp

575 °C (lit.)

density

2.75 g/mL at 25 °C (lit.)

SMILES string

[K+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.K/c1-7(2,3,4,5)6;/q-1;+1

InChI key

YZDGRYDIGCWVND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Multifunctional potassium hexafluorophosphate passivate interface defects for high efficiency perovskite solar cells: Potassium hexafluorophosphate (KPF6) is used to enhance the interface between SnO2 quantum dots and perovskite (Z Wang et al., 2021).
  • Potassium hexafluorophosphate additive enables stable lithium–sulfur batteries: An optimized amount of KPF6 added to the electrolyte improves the cycling stability of lithium-sulfur batteries (J Li et al., 2020).
  • Dual electrolyte additives of potassium hexafluorophosphate and tris (trimethylsilyl) phosphite for anode-free lithium metal batteries: KPF6 and tris(trimethylsilyl) phosphite enhance the electrochemical performance of anode-free lithium metal batteries (TM Hagos et al., 2019).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihua Wang et al.
Frontiers in bioengineering and biotechnology, 8, 655-655 (2020-07-23)
We explore a sandwich-type gold nanoparticle coated reduced graphene oxide (rGO-AuNP) as an effective nanotheranostic platform for the second near-infrared (NIR-II) window photoacoustic (PA) imaging-guided photothermal therapy (PTT) in ovarian cancer. The PEG was loaded onto the AuNPs surface to
Kosuke Doki et al.
Drug metabolism and pharmacokinetics, 30(4), 257-262 (2015-07-22)
The aims of this study were to clarify whether the ratio of S- to R-flecainide (S/R ratio) in the serum flecainide concentration was associated with the stereoselectivity of flecainide metabolism, and to investigate the effects of the cytochrome P450 (CYP)
Anil K Gorle et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16713-16725 (2014-10-02)
A series of inert tri- and tetra-nuclear polypyridylruthenium(II) complexes that are linked by the bis[4(4'-methyl-2,2'-bipyridyl)]-1,n-alkane ligand ("bb(n)" for n = 10, 12 and 16) have been synthesised and their potential as antimicrobial agents examined. Due to the modular nature of
Anil K Gorle et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(29), 10472-10481 (2015-06-05)
Ruthenium(II) complexes containing the tetradentate ligand bis[4(4'-methyl-2,2'-bipyridyl)]-1,n-alkane ("bbn "; n=10 and 12) have been synthesised and their geometric isomers separated. All [Ru(phen)(bbn )](2+) (phen=1,10-phenanthroline) complexes exhibited excellent activity against Gram-positive bacteria, but only the cis-α-[Ru(phen)(bb12 )](2+) species showed good activity
Jeffrey A Therrien et al.
Inorganic chemistry, 53(24), 12962-12972 (2014-10-23)
A series of pyridine- and lutidine-linked bis-N-heterocyclic carbene (NHC) palladium pincer complexes were electrochemically characterized and screened for CO2 reduction capability with 2,2,2-trifluoroethanol, acetic acid, or 2,2,2-trifluoroacetic acid (TFA) as proton sources. The lutidine-linked pincer complexes electrocatalytically reduce CO2 to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service