Skip to Content
Merck
All Photos(2)

Key Documents

227722

Sigma-Aldrich

Silver hexafluorophosphate

98%

Synonym(s):

Silver(I) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgPF6
CAS Number:
Molecular Weight:
252.83
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

core: silver
reagent type: catalyst

mp

102 °C (dec.) (lit.)

SMILES string

[Ag+].F[P-](F)(F)(F)(F)F

InChI

1S/Ag.F6P/c;1-7(2,3,4,5)6/q+1;-1

InChI key

SCQBROMTFBBDHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver hexafluorophosphate can be used as a catalyst for the sulfimidation, hydrogenation of aldehydes to alcohols, and hydroacyloxylation of alkynes with carboxylic acids.

Application

Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials. Preparation of supramolecular networks of Ag(I) complexes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver-Catalyzed Hydrogenation of Aldehydes in Water
Jia Z, et al.
Angewandte Chemie (International Edition in English), 52, 11871-11874 (2013)
Journal of Organometallic Chemistry, 692, 4093-4093 (2007)
Silver-Catalyzed Enantioselective Sulfimidation Mediated by Hydrogen Bonding Interactions
Annapureddy R, et al.
Angewandte Chemie (International Edition in English), 133, 7999-8005 (2021)
Gold (I)-catalyzed addition of carboxylic acids to alkynes
Chary B, et al.
The Journal of Organic Chemistry, 75, 7928-7931 (2010)
Keith A. Hirsch et al.
Inorganic chemistry, 36(14), 2960-2968 (1997-07-02)
Coordination networks of 3,3'-dicyanodiphenylacetylene (3,3'-DCPA, 1) with silver(I) salts characterized by single-crystal X-ray analysis are described. Network topology is found to depend on both the counterion and solvent employed during crystallization. The conformation adopted by the ligand varies between planar

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service