Skip to Content
Merck
All Photos(2)

Key Documents

256005

Sigma-Aldrich

2,6-Pyridinedicarboxaldehyde

97%

Synonym(s):

2,6-Bis(formyl)pyridine, 2,6-Diformylpyridine, 2,6-Pyridinedialdehyde, Pyridine-2,6-dicarbaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$36.40
1 G
$138.00

$36.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$36.40
1 G
$138.00

About This Item

Empirical Formula (Hill Notation):
C7H5NO2
CAS Number:
Molecular Weight:
135.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$36.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

bp

152-154 °C/103 mmHg (lit.)

mp

124-125 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccc(n1)C([H])=O

InChI

1S/C7H5NO2/c9-4-6-2-1-3-7(5-10)8-6/h1-5H

InChI key

PMWXGSWIOOVHEQ-UHFFFAOYSA-N

General description

2,6-Pyridinedicarboxaldehyde can serve as a building block in the synthesis of various organic compounds.[1][2]

Application

2,6-Pyridinedicarboxaldehyde has been used in preparation of:
  • functionalized resin Amberlite XAD-4[2]
  • boron-dipyrromethene (BODIPY)-based fluorescence probe with a N,N′-(pyridine-2, 6-diylbis(methylene))-dianiline substituent[3]
  • novel N-heterocyclic chitosan aerogel derivatives[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cennet Karadaş et al.
Food chemistry, 141(2), 655-661 (2013-06-26)
The synthesis and characterization of the resin Amberlite XAD-4 functionalized with 2,6-pyridinedicarboxaldehyde and its application in an on-line system for the preconcentration of cadmium, cobalt, copper, lead and manganese prior to determination using flame atomic absorption spectrometry (FAAS) is proposed.
Muhammad Saleem et al.
Journal of fluorescence, 26(1), 11-22 (2015-11-21)
Herein, we reported the ditriazole Schiff base derivative 1 and evaluated its photophysical properties on induction of varieties of metal ions including Na(+), Ag(+), Ni(2+), Mn(2+), Pd(2+), Co(2+), Hg(2+), Cu(2+), Pb(2+), Cd(2+), Zn(2+), Sn(2+), Fe(2+), Fe(3+), Cr(3+) and Al(3+), in
Hua Lu et al.
The journal of physical chemistry. A, 113(51), 14081-14086 (2009-12-03)
A boron-dipyrromethene (BODIPY)-based fluorescence probe with a N,N'-(pyridine-2, 6-diylbis(methylene))-dianiline substituent (1) has been prepared by condensation of 2,6-pyridinedicarboxaldehyde with 8-(4-amino)-4,4-difluoro-1,3,5,7-tetramethyl-4-bora-3a,4a-diaza-s-indacene and reduction by NaBH(4). The sensing properties of compound 1 toward various metal ions are investigated via fluorometric titration in
Santosh Kumar et al.
International journal of biological macromolecules, 45(4), 330-337 (2009-08-12)
The novel N-heterocyclic chitosan aerogel derivatives were prepared by reacting 79% deacetylated chitosan separately with 4-pyridinecarboxaldehyde and 2,6-pyridinedicarboxaldehyde followed by subsequent solvent exchange into acetone, filteration and lyophilization. The identity of the Schiff bases was confirmed by UV-vis absorption spectroscopy
Muhammad Hanif et al.
Molecules (Basel, Switzerland), 24(2) (2019-01-19)
The present study focuses on the design and synthesis of a cage-like organic skeleton containing two triazole rings jointed via imine linkage. These molecules can act as urease inhibitors. The in-vitro urease inhibition screening results showed that the combination of

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service