Skip to Content
Merck
All Photos(3)

Key Documents

154369

Sigma-Aldrich

2,6-Pyridinedimethanol

98%

Synonym(s):

2,6-Bis(hydroxymethyl)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$49.60
25 G
$170.00
100 G
$602.00

$49.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$49.60
25 G
$170.00
100 G
$602.00

About This Item

Empirical Formula (Hill Notation):
C7H9NO2
CAS Number:
Molecular Weight:
139.15
Beilstein:
116016
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$49.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

112-114 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1cccc(CO)n1

InChI

1S/C7H9NO2/c9-4-6-2-1-3-7(5-10)8-6/h1-3,9-10H,4-5H2

InChI key

WWFMINHWJYHXHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Pyridinedimethanol can be used as a ligand to synthesize a variety of metal complexes[1] and catalysts.[2] It can also be used as a reactant to prepare Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and [Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)).[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jui-Ting Sun et al.
Polymers, 12(11) (2020-11-21)
In this study, the researchers successfully embellished the surface of halloysite (Ag/HNTs) with silver using halloysite, silver nitrate (AgNO3), and polyvinylpyrrolidone (PVP). The researchers then prepared polyurethane that contained pyridine ring by using 4,4'-diphenylmethane diisocyanate (MDI) and polytetramethylene glycol (PTMG)
Large bite bisphosphite, 2,6-C5H3N{CH2OP(?OC10H6)(?-S)(C10H6O?)}2: Synthesis, derivatization, transition metal chemistry and application towards hydrogenation of olefins
Benudhar P, Maravanji SB
Journal of Organometallic Chemistry, 692, 1683-1689 (2007)
Homogeneous and heterogeneous ruthenium catalysts in the synthesis of fine chemicals
Wolfgang M, et al.
Catalysis Today, 121, 1-2, 140-150 (2007)
Taketo Taguchi et al.
Dalton transactions (Cambridge, England : 2003), 39(38), 9131-9139 (2010-09-08)
The syntheses, crystal structures and magnetochemical characterization are reported for two new Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) (3) and [Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)) (4). They were synthesized from the use of the potentially O,N,O tridentate chelate, 2,6-pyridinedimethanol (pdmH(2)), in the presence or absence of carboxylate
Hao Xu et al.
Science and technology of advanced materials, 21(1), 450-460 (2020-09-18)
We report the dynamic behavior of diselenide-containing hydrophilic polyurethanes and hydrogels based on diselenide exchange reactions in an aqueous media. Diselenide-containing linear and cross-linked polyurethanes were synthesized via polyaddition reactions using diselenide-containing diol in combination with pyridinium diol that enhances

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service