Skip to Content
Merck
All Photos(1)

Key Documents

178322

Sigma-Aldrich

Sodium dicyanamide

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$72.80
100 G
$277.00

$72.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$72.80
100 G
$277.00

About This Item

Linear Formula:
NaN(CN)2
CAS Number:
Molecular Weight:
89.03
Beilstein:
3913818
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

$72.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

form

powder

impurities

<3% NaCl

functional group

amine

SMILES string

[Na]N(C#N)C#N

InChI

1S/C2N3.Na/c3-1-5-2-4;/q-1;+1

InChI key

IXBPPZBJIFNGJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Nickel diacetylpyridine dioximate pentanuclear, octanuclear and decanuclear cluster complexes
  • Decanuclear manganese(II/III) oxo azide diolate clusters as supertetrahedral building-blocks to design extended magnetic networks
  • Ruthenium bipyridine complexes entrapped in iron and manganese dicyanamide two-dimensional anionic sheet-like coordination polymeric networks exhibiting dual emission photoluminescence
  • Lead dicyanamide-bridged nitronyl nitroxide radical polymeric complex
  • Antiferromagnetically coupled hexanuclear copper(II) Schiff base complexes with dicyanamido bridges
  • CuII2L based polymeric ladders using dicyanamide bridges
  • Luminous hexacoordinated coordination polymers of cadmium(II), containing dicyanamide bridges

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danah Al-Masri et al.
Physical chemistry chemical physics : PCCP, 22(32), 18102-18113 (2020-08-08)
Ionic liquids and plastic crystals based on pyrrolidinium cations are recognised for their advantageous properties such as high conductivity, low viscosity, and good electrochemical and thermal stability. The pyrrolidinium ring can be substituted with symmetric or asymmetric alkyl chain substituents
Bing-Jyun Lu et al.
Scientific reports, 9(1), 13253-13253 (2019-09-15)
Wood has been a crucial natural material for human civilization since prehistoric times. In archaeology, the examination of the wood microstructure is important for the study of architecture, musical instruments, sculptures, and so on. Scanning electron microscopy (SEM) examination is

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service