Skip to Content
Merck
All Photos(1)

Key Documents

360899

Sigma-Aldrich

Ethyl potassium malonate

98%

Synonym(s):

Monoethyl malonate potassium salt, Potassium monoethyl malonate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$111.00
100 G
$401.00

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$111.00
100 G
$401.00

About This Item

Linear Formula:
C2H5OCOCH2CO2K
CAS Number:
Molecular Weight:
170.20
Beilstein:
3721682
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

194 °C (dec.) (lit.)

functional group

ester

SMILES string

[K+].CCOC(=O)CC([O-])=O

InChI

1S/C5H8O4.K/c1-2-9-5(8)3-4(6)7;/h2-3H2,1H3,(H,6,7);/q;+1/p-1

InChI key

WVUCPRGADMCTBN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl potassium malonate (potassium ethyl malonate) reacts with aryl nitriles in the presence of zinc chloride and a catalytic amount of Hünig′s base to yield β-amino acrylates.[1] Ethyl potassium malonate is formed as an intermediate during the synthesis of ethyl tert-butyl malonate.[2]

Application

Ethyl potassium malonate (potassium ethyl malonate) may be used to generate (trimethylsilyl)ethyl malonate in situ, which can be acylated to prepare a variety of β-ketoesters or alkylidene malonates.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Hoon Lee et al.
The Journal of organic chemistry, 72(26), 10261-10263 (2007-12-01)
Reaction of aryl nitriles with potassium ethyl malonate in the presence of zinc chloride and a catalytic amount of Hünig's base provided beta-amino acrylates in moderate to good yield. Compared to the classical Blaise reaction, this reaction is safer (endothermic)
Ethyl tert-Butyl Malonate.
Strube RE.
Organometallic Syntheses, 34-34 (1963)
A process for the synthesis of ?-ketoesters using in-situ generated (trimethylsilyl) malonates
Wang X, et al.
Tetrahedron Letters, 35(50), 9323-9326 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service