Skip to Content
Merck
All Photos(1)

Key Documents

151440

Sigma-Aldrich

Methyl chlorooxoacetate

96%

Synonym(s):

Monomethyl oxalyl chloride, Chloroglyoxylic acid methyl ester, Methyl (chloroformyl)formate, Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxalate, Methyl oxalyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOOCH3
CAS Number:
Molecular Weight:
122.51
Beilstein:
1071541
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Quality Level

Assay

96%

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.419 (lit.)

bp

118-120 °C (lit.)

density

1.332 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

SMILES string

COC(=O)C(Cl)=O

InChI

1S/C3H3ClO3/c1-7-3(6)2(4)5/h1H3

InChI key

ZXUQEPZWVQIOJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Regioselective synthesis of fused coumarins
  • Cyclization to form substituted isoxazoles and heterocycles
  • Intramolecular Wittig reactions
  • Silyl enol ether acylation
  • Iron-mediated cleavage of C-C bonds

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service