Skip to Content
Merck
All Photos(2)

Key Documents

142875

Sigma-Aldrich

2,6-Pyridinedicarbonyl dichloride

97%

Synonym(s):

2,6-Pyridinedicarbonyl chloride, Pyridine-2,6-dicarboxylic acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$93.80
25 G
$251.00

$93.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$93.80
25 G
$251.00

About This Item

Empirical Formula (Hill Notation):
C7H3Cl2NO2
CAS Number:
Molecular Weight:
204.01
Beilstein:
131556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$93.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

284 °C (lit.)

mp

56-58 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)c1cccc(n1)C(Cl)=O

InChI

1S/C7H3Cl2NO2/c8-6(11)4-2-1-3-5(10-4)7(9)12/h1-3H

InChI key

GWHOGODUVLQCEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Pyridinedicarbonyl dichloride can be used as a starting material to synthesize:
  • Pyridine-based polyamido-polyester optically active macrocycles by reacting with chiral diamine dihydrobromides.[1]
  • Pyridine-bridged 2,6-bis-carboxamide Schiff′s bases by treating with L-alanine or 2-methyl-alanine methyl esters.[2]
  • N,N′-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide by reacting with 1-pyrenemethylamine hydrochloride in the presence of a base.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of some new pyridine-2, 6-carboxamide-derived Schiff bases as potential antimicrobial agents
Al-Omar MA and Amr Abd El-Galil E
Molecules (Basel), 15(7), 4711-4721 (2010)
Mohamed A Al-Omar et al.
Molecules (Basel, Switzerland), 15(7), 4711-4721 (2010-07-27)
A series of pyridine-bridged 2,6-bis-carboxamide Schiff's bases has been prepared starting from 2,6-pyridinedicarbonyl dichloride (1) and L-alanine or 2-methyl-alanine methyl ester.The coupling of acid chloride 1 with L-alanine methyl ester hydrochloride -or 2-methylalanine methyl ester hydrochloride gave the corresponding 2,6-bis-carboxamide
H Zhao et al.
The Journal of organic chemistry, 65(10), 2933-2938 (2000-05-18)
Five new chiral macrocycles, 3a-e, have been prepared by the acylation cyclization of chiral diamine dihydrobromide intermediates 2a-c with 2,6-pyridinedicarbonyl dichloride in highly diluted solution at room temperature. The chiral diesters 1a-c needed for the preparation of the macrocycles were
Recognition of sequence-information in synthetic copolymer chains by a conformationally-constrained tweezer molecule
Colquhoun HM, et al.
Faraday Discussions, 143(7), 205-220 (2009)
Synthesis and characterization of pyridine-based polyamido-polyester optically active macrocycles and enantiomeric recognition for D-and L-amino acid methyl ester hydrochloride
Zhao H and Hua W
The Journal of Organic Chemistry, 65(10), 2933-2938 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service