Skip to Content
Merck
All Photos(1)

Key Documents

544655

Sigma-Aldrich

4-Methoxybenzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$103.00

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$103.00

About This Item

Linear Formula:
CH3OC6H4SO2NH2
CAS Number:
Molecular Weight:
187.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

111-115 °C (lit.)

SMILES string

COc1ccc(cc1)S(N)(=O)=O

InChI

1S/C7H9NO3S/c1-11-6-2-4-7(5-3-6)12(8,9)10/h2-5H,1H3,(H2,8,9,10)

InChI key

MSFQEZBRFPAFEX-UHFFFAOYSA-N

Related Categories

General description

4-Methoxybenzenesulfonamide, also known as p-methoxybenzenesulfonamide, is an aromatic sulfonamide. It can undergo polyaddition with vinyloxiranes in the presence of a palladium catalyst.[1]

Application

4-Methoxybenzenesulfonamide may be used in the preparation of S,S-dimethyl-N-p-methoxybenzenesulfonylsulfilimine.[1] It may also be used as a ligand to generate novel bis-tetrahydrofuran–based human immunodeficiency virus (HIV) protease inhibitor.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbodiimide-sulfoxide reactions. XII. Reactions of sulfonamides.
The Journal of Organic Chemistry, 36(24), 3686-3691 (1971)
Palladium (0)-catalyzed polyaddition of bifunctional vinyloxirane with nitrogen nucleophiles. Synthesis of polymers containing an allylamine moiety in the main chain and pendant hydroxyl groups.
Koizumi T, et al.
Macromolecules, 36(15), 5882-5884 (2003)
Bis-tetrahydrofuran: a privileged ligand for darunavir and a new generation of hiv protease inhibitors that combat drug resistance.
Arun K Ghosh et al.
ChemMedChem, 1(9), 939-950 (2006-08-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service