Skip to Content
Merck
All Photos(2)

Key Documents

121193

Sigma-Aldrich

4,4′-Trimethylenedipyridine

98%

Synonym(s):

1,3-Bis(4-pyridyl)propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

57-60 °C (lit.)

SMILES string

C(Cc1ccncc1)Cc2ccncc2

InChI

1S/C13H14N2/c1(2-12-4-8-14-9-5-12)3-13-6-10-15-11-7-13/h4-11H,1-3H2

InChI key

OGNCVVRIKNGJHQ-UHFFFAOYSA-N

Application

4,4′-Trimethylenedipyridine was used as ligand to study the solvent effects on the formal potential of redox-active self-assembling system [Os(bpy)2(dipy)Cl]1+ (bpy= 2,2′-bipyridine, dipy= 4,4′-Trimethylenedipyridine) in different organic solvents and aqueous solutions[1]. It was used in the synthesis of two- and three-dimensional cadmium-organic frameworks[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron-transfer study and solvent effects on the formal potential of a redox-active self-assembling monolayer.
Acevedo D and Abruna HD.
The Journal of Physical Chemistry, 95(23), 9590-9594 (1991)
Filipe A Almeida Paz et al.
Inorganic chemistry, 43(13), 3882-3893 (2004-06-23)
Three novel cadmium-organic frameworks built-up from 1,3,5-benzenetricarboxylate anions (HXBTC(x-3)) and 4,4'-trimethylenedipyridine (TMD) have been hydrothermally synthesized, and characterized using single-crystal X-ray diffraction, thermoanalytical measurements, elemental analysis, and IR and Raman spectroscopies: [Cd(HBTC)(TMD)(2)].8.5H(2)O (I), [Cd(HBTC)(TMD)(H(2)O)].4.5H(2)O (II), and [Cd(2)(BTC)(TMD)(2)(NO(3))].3H(2)O (III), with structures
Xueling Shan et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(6), 639-644 (2019-02-13)
A novel electrochemiluminescence (ECL) sensor for detection of brilliant blue FCF (BB) has been developed using Ru@Zn-MOF/nafion modified GCE (glass carbon electrode) in this research. Different from conventional method for usage of Ru(bpy)32+ in solution-phase, Ru(bpy)32+ here was immobilized on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service