Skip to Content
Merck
All Photos(2)

Key Documents

192260

Sigma-Aldrich

4,4′-Trimethylenebis(1-methylpiperidine)

≥98%

Synonym(s):

4,4′-(1,3-Propanediyl)bis[1-methyl-piperidine]

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$225.00

About This Item

Empirical Formula (Hill Notation):
C15H30N2
CAS Number:
Molecular Weight:
238.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

refractive index

n20/D 1.482 (lit.)

bp

215 °C/50 mmHg (lit.)

mp

13 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

SMILES string

CN1CCC(CCCC2CCN(C)CC2)CC1

InChI

1S/C15H30N2/c1-16-10-6-14(7-11-16)4-3-5-15-8-12-17(2)13-9-15/h14-15H,3-13H2,1-2H3

InChI key

JWOTWWORMYMZCR-UHFFFAOYSA-N

Application

4,4′-Trimethylenebis(1-methylpiperidine) has been used in the synthesis of:
  • di-, tri- and tetracationic monomeric and homodimeric monomethine cyanine dyes for nucleic acid detection[1]
  • structure directing agents containing 4,4′-trimethylenebis(1-methylpiperidine)[2]
  • piperidine-based dicationic ionic liquids[3]
Reactant for synthesis of cyanine dyes for nucleic acid detection

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of hierarchical Beta using piperidine based multi-ammonium surfactants.
Kore RR, et al.
Royal Society of Chemistry Advances, 3(5), 1317-1322 (2013)
Rajkumar Kore et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(51), 14360-14365 (2011-11-25)
Piperidine- and imidazole-based dicatoinic ionic liquids have been developed for the synthesis of zeolite Beta. Hierarchical Beta has a larger surface area and pore volume than conventional Beta. Beta derived from a dicationic ionic liquid exhibited remarkably higher catalytic activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service