Skip to Content
Merck
All Photos(2)

Key Documents

143057

Sigma-Aldrich

Aldrithiol-4

98%

Synonym(s):

4,4′-Dipyridyl disulfide, 4,4′-Dithiodipyridine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$100.00
5 G
$272.00
25 G
$1,310.00

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$100.00
5 G
$272.00
25 G
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C10H8N2S2
CAS Number:
Molecular Weight:
220.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

76-78 °C (lit.)

solubility

95% ethanol: soluble 5%, clear, colorless to light yellow

functional group

disulfide

storage temp.

2-8°C

SMILES string

S(Sc1ccncc1)c2ccncc2

InChI

1S/C10H8N2S2/c1-5-11-6-2-9(1)13-14-10-3-7-12-8-4-10/h1-8H

InChI key

UHBAPGWWRFVTFS-UHFFFAOYSA-N

Application

  • Aldrithiol-4 was used as cosubstrate in a study to develop highly specific inhibitors against mannose-binding lectin-associated serine proteases by in vitro evolution technology:phage display[1].
  • It was used to study the O2 equilibria of recombinant human neuroglobin and cytoglobin measured under close to physiological conditions[2].
  • It was used to modify gold surfaces for protein studies[3].

Legal Information

Aldrithiol is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shwu-Jiuan Sheu et al.
Investigative ophthalmology & visual science, 44(3), 1237-1244 (2003-02-26)
To identify the mechanisms by which oxidative stress with oxidizing agents alters the activity of ion channels in human retinal pigment epithelial (RPE) cells. The effects of oxidizing agents on ion currents were investigated in human RPE R-50 cells with
Nikitas Bistolas et al.
Biochemical and biophysical research communications, 314(3), 810-816 (2004-01-27)
The spectroelectrochemistry of camphor-bound cytochrome P450cam (P450cam) using gold electrodes is described. The electrodes were modified with either 4,4(')-dithiodipyridin or sodium dithionite. Electrolysis of P450cam was carried out when the enzyme was in solution, while at the same time UV-visible
Iveta Imriskova-Sosova et al.
Biochemistry, 44(51), 16949-16956 (2005-12-21)
Members of the Sco protein family are implicated in the assembly of the respiratory complex cytochrome c oxidase. Several possible roles have been proposed for Sco: a copper delivery agent, a site-specific thiol reductase, and an indicator of cellular redox
Soong-Yu Kuo et al.
Archives of toxicology, 77(11), 630-637 (2003-08-21)
The effect of oxidants on Ca(2+) movement in osteoblasts is unclear. In this study, we show that 4,4'-dithiodipyridine (4,4'-DTDP), a reactive disulphide that mobilizes Ca(2+) in muscle, induces an increase in cytoplasmic free-Ca(2+) concentrations ([Ca(2+)](i)) in MG63 human osteosarcoma cells
H Reynaldo López-Mirabal et al.
FEMS yeast research, 7(3), 391-403 (2007-01-27)
Dipyridyl disulfide (DPS) is a highly reactive thiol oxidant that functions as electron acceptor in thiol-disulfide exchange reactions. DPS is very toxic to yeasts, impairing growth at low micromolar concentrations. The genes TRX2 (thioredoxin), SOD1 (superoxide dismutase), GSH1 (gamma-glutamyl-cysteine synthetase)

Articles

Clostridium perfringens can cause contamination in undercooked or improperly sterilized canned foods and water. Learn to detect, identify, and differentiate this pathogen.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service