Skip to Content
Merck
All Photos(2)

Documents

245739

Sigma-Aldrich

4,4′-Dimethyl-2,2′-dipyridyl

99%

Synonym(s):

2,2′-Bi(γ-picoline), 4,4′-Dimethyl-2,2′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
128995
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

169-174 °C (lit.)

SMILES string

Cc1ccnc(c1)-c2cc(C)ccn2

InChI

1S/C12H12N2/c1-9-3-5-13-11(7-9)12-8-10(2)4-6-14-12/h3-8H,1-2H3

InChI key

NBPGPQJFYXNFKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Dimethyl-2,2′-dipyridyl was used in the synthesis of a series of o-phenanthroline-substituted ruthenium(II) complexes. It was also used as a starting material for the synthesis of a blue luminescent molecule with emission at 450nm and a quantum yield of 43%.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Materials Chemistry, 16, 4468-4468 (2006)
Ricardo Alvarez et al.
Dalton transactions (Cambridge, England : 2003), 49(28), 9632-9640 (2020-07-03)
The Nano-Crystallization method has been extensively tested for the growth of single crystals of cationic coordination compounds, which are soluble in water. All three studied, diverse metal complexes could be crystallized with the help of pipetting robots commonly used for
Byeong Hyo Kim et al.
Talanta, 62(3), 595-602 (2008-10-31)
A series of o-phenanthroline-substituted ruthenium(II) complexes containing 2,2'-dipyridyl, 2-(2-pyridyl)benzimidazole, 2-(2-pyridyl)-N-methylbenzimidazole, 4-carboxymethyl-4'-methyl-2,2'-dipyridyl, and/or 4,4'-dimethyl-2,2'-dipyridyl ligands were synthesized and examined as potent electrochemiluminescent (ECL) materials. The characteristics of these complexes, regarding their electrochemical redox potentials and relative ECL intensities for tripropylamine were
Maxime Mourer et al.
Bioorganic & medicinal chemistry, 20(6), 2035-2041 (2012-03-01)
Various polycharged calix[4]arenes were assayed as anti-mycobacterial agents against Mycobacterium tuberculosis, H(37)Rv strain. The sulfonate, carboxylate and phosphonate anionic species displayed no activity. Cationic derivatives integrating four aminoethyl groups at the upper rim and two 6,6'-dimethyl-2,2'-bipyridyl- or 4,4'-dimethyl-2,2'-bithiazolyl subunits at
Yang-Hui Luo et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16937-16942 (2014-10-11)
A new iron(II) complex based on the 4,4'-dimethyl-2,2'-bipyridine ligand [Fe(4,4'-dmbpy)3(ClO4)(SCN)·3H2O (1·3H2O)] has been prepared and characterized. Structural studies and Hirshfeld surface analysis for complex 1·3H2O at three different temperatures (300, 240 and 130 K) are described. The UV-vis absorption spectrum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service