Merck
  • Galactosylsphingosine (psychosine)-induced demyelination is attenuated by sphingosine 1-phosphate signalling.
Galactosylsphingosine (psychosine)-induced demyelination is attenuated by sphingosine 1-phosphate signalling.
Journal of cell science (2015-09-12)
Catherine O'Sullivan, Kumlesh K Dev
ABSTRACT
Globoid cell leukodystrophy (Krabbe disease) is a rare infantile neurodegenerative disorder. Krabbe disease is caused by deficiency in the lysosomal enzyme galactocerebrosidase (GALC) resulting in accumulation, in the micromolar range, of the toxic metabolite galactosylsphingosine (psychosine) in the brain. Here we find that psychosine induces human astrocyte cell death probably via an apoptotic process in a concentration- and time-dependent manner (EC50 ∼ 15 μM at 4 h). We show these effects of psychosine are attenuated by pre-treatment with the sphingosine 1-phosphate receptor agonist pFTY720 (fingolimod) (IC50 ∼ 100 nM). Psychosine (1 μM, 10 μM) also enhances LPS-induced (EC50 ∼ 100 ng/ml) production of pro-inflammatory cytokines in mouse astrocytes, which is also attenuated by pFTY720 (1 μM). Most notably, for the first time, we show that psychosine, at a concentration found in the brains of patients with Krabbe disease (EC50 ∼ 100 nM), directly induces demyelination in mouse organotypic cerebellar slices in a manner that is independent of pro-inflammatory cytokine response and that pFTY720 (0.1 nM) significantly inhibits. These results support the idea that psychosine is a pathogenic agent in Krabbe disease and suggest that sphingosine 1-phosphate signalling could be a potential drug target for this disorder.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Anti-Myelin Proteolipid Protein Antibody, CT, clone PLPC1, clone PLPC1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse
Avanti
Galactosyl(β) Sphingosine-d5, Avanti Research - A Croda Brand 860637P, powder