Skip to Content
Merck
All Photos(5)

Key Documents

M5655

Sigma-Aldrich

Thiazolyl Blue Tetrazolium Bromide

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)

Synonym(s):

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide, MTT, Methylthiazolyldiphenyl-tetrazolium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16BrN5S
CAS Number:
Molecular Weight:
414.32
Beilstein:
4081397
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

Assay

≥97.5% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

mp

195 °C (dec.) (lit.)

solubility

H2O: 5 mg/mL

storage temp.

2-8°C

SMILES string

[Br-].Cc1nc(sc1C)-[n+]2nc(nn2-c3ccccc3)-c4ccccc4

InChI

1S/C18H16N5S.BrH/c1-13-14(2)24-18(19-13)23-21-17(15-9-5-3-6-10-15)20-22(23)16-11-7-4-8-12-16;/h3-12H,1-2H3;1H/q+1;/p-1

InChI key

AZKSAVLVSZKNRD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.
Thiazolyl Blue Tetrazolium Bromide has been used as a colorimetric metabolic activity indicator in cell viability assays (MTT assay). It has also been used to determine cell proliferation.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication, mechanical properties, and biocompatibility of graphene-reinforced chitosan composites
Fan H, et al.
Biomacromolecules, 11(9), 2345-2351 (2010)
Nanoparticles made from novel starch derivatives for transdermal drug delivery
Santander Ortega MJ, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 141(1), 85-92 (2010)
UNC569, a novel small-molecule mer inhibitor with efficacy against acute lymphoblastic leukemia in vitro and in vivo
Christoph S, et al.
Molecular Cancer Therapeutics (2013)
Sahar Yarahmadi et al.
Gene, 711, 143939-143939 (2019-06-21)
Sirtuin 1 is one of the regulators of cell growth and survival and its inhibition is suggested as a suitable mechanism to overcome breast cancer development. In this study we explored the role of miR-211-5p in SIRT1/p53 pathway and its
Dasatinib synergizes with doxorubicin to block growth, migration, and invasion of breast cancer cells
Pichot CS, et al.
British Journal of Cancer, 101(1), 38-38 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service