Skip to Content
Merck
All Photos(1)

Key Documents

49139

Sigma-Aldrich

D-(+)-Glucose

BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

grade

anhydrous

Quality Level

product line

BioUltra

Assay

≥99.5% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]20/D +53±2°, 3 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

pH

5.0-7.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.10
λ: 280 nm Amax: 0.10

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Neutralizes growth inhibition by myxothiazol in Saccharomyces cerevisiae, Mucor hiemalis and Candida albicans; Component of the liquid stationary phase in the GLC separation of nitroxylene and xylenol isomers

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Thierbach et al.
Antimicrobial agents and chemotherapy, 19(4), 504-507 (1981-04-01)
Myxothiazol, a new antibiotic from the myxobacterium Myxococcus fulvus, inhibited the growth of many yeasts and fungi at concentrations between 0.01 and 3 micrograms/ml. It was generally inactive against bacteria. The inhibitory effect was cytostatic. With Candida albicans, Saccharomyces cerevisiae
Separation behaviour of some isomeric organic compounds on sugars, sugar alcohols and their mixed phases by gas-liquid chromatography.
Ono, A.
Journal of Chromatography A, 197, 251-251 (1980)
Jashanpreet Kaur et al.
PloS one, 10(3), e0122354-e0122354 (2015-03-27)
Genotyping studies of Australian Scedosporium isolates have revealed the strong prevalence of a recently described species: Scedosporium aurantiacum. In addition to occurring in the environment, this fungus is also known to colonise the respiratory tracts of cystic fibrosis (CF) patients.
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell
Lars P Bechmann et al.
Journal of hepatology, 56(4), 952-964 (2011-12-17)
It is widely known that the liver is a central organ in lipogenesis, gluconeogenesis and cholesterol metabolism. However, over the last decades, a variety of pathological conditions highlighted the importance of metabolic functions within the diseased liver. As observed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service